Product Details

SNP ID
rs79489583
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:117917504 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCAGGACCCAGCTATCCTGAAAA[C/T]ATTAAGGGCTACGAGCTAGAACCAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC93 PubMed Links

Gene Details

Gene
CCDC93
Gene Name
coiled-coil domain containing 93
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019044.4 4903 UTR 3 NP_061917.3
XM_006712600.2 4903 Intron XP_006712663.1
XM_011511359.1 4903 UTR 3 XP_011509661.1
XM_011511361.1 4903 UTR 3 XP_011509663.1
XM_011511362.1 4903 UTR 3 XP_011509664.1
XM_011511363.2 4903 Intron XP_011509665.1
Gene
LOC101929908
Gene Name
uncharacterized LOC101929908
There are no transcripts associated with this gene.

View Full Product Details