Product Details

SNP ID
rs76884980
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:109131068 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGCTTTAGAAACACAACTTTATT[C/T]CTCTACTTTTGACCATTTAAGGAGT
Phenotype
MIM: 601181
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RANBP2 PubMed Links

Gene Details

Gene
RANBP2
Gene Name
RAN binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006267.4 Intron NP_006258.3
XM_005264002.2 Intron XP_005264059.1
XM_005264003.2 Intron XP_005264060.1
XM_005264004.2 Intron XP_005264061.1
XM_005264005.4 Intron XP_005264062.1
XM_005264007.2 Intron XP_005264064.1
XM_011511575.2 Intron XP_011509877.1
XM_011511576.2 Intron XP_011509878.1
XM_011511578.2 Intron XP_011509880.1
XM_017004623.1 Intron XP_016860112.1
XM_017004624.1 Intron XP_016860113.1
XM_017004625.1 Intron XP_016860114.1
Gene
SH3RF3
Gene Name
SH3 domain containing ring finger 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099289.2 Intron NP_001092759.1
XM_011511109.2 Intron XP_011509411.1
XM_011511110.2 Intron XP_011509412.1
Gene
SH3RF3-AS1
Gene Name
SH3RF3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details