Product Details

SNP ID
rs75604464
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:95149721 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCATTCATAGGGCTTTTCTCCA[G/T]TATGAGTTCTTTGATGTTTAATAAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF514 PubMed Links

Gene Details

Gene
ZNF514
Gene Name
zinc finger protein 514
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318005.1 1585 Missense Mutation AAT,ACT N328T NP_001304934.1
NM_032788.2 1585 Missense Mutation AAT,ACT N255T NP_116177.1
XM_006712806.2 1585 Missense Mutation AAT,ACT N255T XP_006712869.1

View Full Product Details