Product Details

SNP ID
rs76935572
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:101261422 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGCCCTTCATTGCTGCTGTGGT[A/G]CCTTCACACTGTGGAGTACCAAGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNOT11 PubMed Links

Gene Details

Gene
CNOT11
Gene Name
CCR4-NOT transcription complex subunit 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017546.4 Intron NP_060016.3
Gene
RNF149
Gene Name
ring finger protein 149
There are no transcripts associated with this gene.

View Full Product Details