Product Details

SNP ID
rs79763068
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:102421153 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGAAAGACAAACTCAGTTCCTG[C/T]TGGGATGGAACTTATATTCTAGTTG
Phenotype
MIM: 604509
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL18RAP PubMed Links

Gene Details

Gene
IL18RAP
Gene Name
interleukin 18 receptor accessory protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003853.3 Intron NP_003844.1
XM_011512087.2 Intron XP_011510389.1
XM_011512088.2 Intron XP_011510390.1
XM_017005173.1 Intron XP_016860662.1

View Full Product Details