Product Details
- SNP ID
-
rs78734289
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:38141442 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTCCTAGATCTATATATGAAATAT[A/G]TAGAAGCCCACCTTCAATCCTCCTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CYP1B1-AS1
PubMed Links
Gene Details
- Gene
- CYP1B1-AS1
- Gene Name
- CYP1B1 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details