Product Details

SNP ID
rs75946468
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:23928715 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAAATGACCAAATGGCATCAAAC[G/T]TAGAGCATTCTGGCCATCTAAGCTC
Phenotype
MIM: 615347
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ATAD2B PubMed Links

Gene Details

Gene
ATAD2B
Gene Name
ATPase family, AAA domain containing 2B
There are no transcripts associated with this gene.

Gene
UBXN2A
Gene Name
UBX domain protein 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181713.3 Intron NP_859064.2
XM_005264166.4 Intron XP_005264223.1
XM_005264167.4 Intron XP_005264224.1
XM_005264168.4 Intron XP_005264225.1
XM_005264169.4 Intron XP_005264226.1
XM_011532633.2 Intron XP_011530935.1
XM_017003517.1 Intron XP_016859006.1
XM_017003518.1 Intron XP_016859007.1

View Full Product Details