Product Details
- SNP ID
-
rs75201250
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:24204806 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCATTTTAGTGGCACTGGACACAC[C/T]GTTAGAAAGCATTCCTTTATTCAGT
- Phenotype
-
MIM: 604464
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ITSN2
PubMed Links
Gene Details
- Gene
- ITSN2
- Gene Name
- intersectin 2
View Full Product Details