Product Details

SNP ID
rs78351099
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74561016 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGCAAATATAGAGAATCCAGATC[C/T]CAGGAAGAGGAAAAGGAGAAGGAGG
Phenotype
MIM: 602919 MIM: 607163
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DOK1 PubMed Links

Gene Details

Gene
DOK1
Gene Name
docking protein 1
There are no transcripts associated with this gene.

Gene
LOXL3
Gene Name
lysyl oxidase like 3
There are no transcripts associated with this gene.

Gene
M1AP
Gene Name
meiosis 1 associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281295.1 Intron NP_001268224.1
NM_001281296.1 Intron NP_001268225.1
NM_001321739.1 Intron NP_001308668.1
NM_138804.4 Intron NP_620159.2
XM_005264152.3 Intron XP_005264209.1
XM_006711946.3 Intron XP_006712009.1
XM_011532548.2 Intron XP_011530850.1
XM_011532549.2 Intron XP_011530851.1
XM_011532550.2 Intron XP_011530852.1
XM_011532551.2 Intron XP_011530853.1
XM_011532552.2 Intron XP_011530854.1
XM_011532553.2 Intron XP_011530855.1
XM_011532554.2 Intron XP_011530856.1

View Full Product Details