Product Details

SNP ID
rs76958153
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:28493124 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTTAATCCACGTGGGCTCTGTGC[C/T]AGGAGCTTAGTCTAGCACCTCCTTG
Phenotype
MIM: 610179
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLB1 PubMed Links

Gene Details

Gene
PLB1
Gene Name
phospholipase B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170585.1 Intron NP_001164056.1
NM_153021.4 Intron NP_694566.4
XM_011532579.2 Intron XP_011530881.1
XM_011532581.2 Intron XP_011530883.1
XM_011532584.2 Intron XP_011530886.1
XM_011532588.2 Intron XP_011530890.1
XM_011532589.2 Intron XP_011530891.1
XM_011532590.2 Intron XP_011530892.1
XM_011532591.2 Intron XP_011530893.1
XM_011532593.2 Intron XP_011530895.1
XM_011532601.2 Intron XP_011530903.1
XM_011532602.2 Intron XP_011530904.1
XM_011532603.2 Intron XP_011530905.1
XM_011532606.2 Intron XP_011530908.1
XM_011532607.2 Intron XP_011530909.1
XM_011532608.2 Intron XP_011530910.1
XM_011532609.2 Intron XP_011530911.1
XM_011532610.2 Intron XP_011530912.1
XM_017003432.1 Intron XP_016858921.1
XM_017003433.1 Intron XP_016858922.1
XM_017003434.1 Intron XP_016858923.1

View Full Product Details