Product Details

SNP ID
rs76494637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:28761831 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGATTTAACACTGTTGTATGCTTG[C/T]TTTTGGCCAGGAACTATAGGGCAGT
Phenotype
MIM: 600590
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPP1CB PubMed Links

Gene Details

Gene
PPP1CB
Gene Name
protein phosphatase 1 catalytic subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002709.2 Intron NP_002700.1
NM_206876.1 Intron NP_996759.1

View Full Product Details