Product Details

SNP ID
rs74496576
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:12559196 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGACTGCCTTGCCAAATAAAAAAC[A/G]TAACAGGTGTTTTAAGTGGGAAAAA
Phenotype
MIM: 608426
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MKRN2 PubMed Links

Gene Details

Gene
MKRN2
Gene Name
makorin ring finger protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271707.1 Intron NP_001258636.1
NM_014160.4 Intron NP_054879.3
Gene
MKRN2OS
Gene Name
MKRN2 opposite strand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195279.1 Intron NP_001182208.1
XM_006712909.3 Intron XP_006712972.1
XM_011533241.2 Intron XP_011531543.1
XM_011533242.2 Intron XP_011531544.1
XM_011533243.2 Intron XP_011531545.1

View Full Product Details