Product Details

SNP ID
rs77443885
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:13483910 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGCCCCTGTCCTCCAACCATAC[A/G]TAGCTCTTTCACTTTGGTCTATTTT
Phenotype
MIM: 607226
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HDAC11 PubMed Links

Gene Details

Gene
HDAC11
Gene Name
histone deacetylase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136041.2 Intron NP_001129513.1
NM_024827.3 Intron NP_079103.2
XM_005265491.1 Intron XP_005265548.1
XM_005265492.1 Intron XP_005265549.1
XM_006713338.1 Intron XP_006713401.1
XM_011534131.1 Intron XP_011532433.1
XM_011534132.2 Intron XP_011532434.2
XM_011534135.1 Intron XP_011532437.1
XM_017007234.1 Intron XP_016862723.1
Gene
HDAC11-AS1
Gene Name
HDAC11 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details