Product Details

SNP ID
rs78242467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:20162174 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCTGTCTCCTGAATGTTGACTGA[A/G]TACTAATCCATGCATGTGTAGGATG
Phenotype
MIM: 602303 MIM: 609168
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KAT2B PubMed Links

Gene Details

Gene
KAT2B
Gene Name
lysine acetyltransferase 2B
There are no transcripts associated with this gene.

Gene
SGO1
Gene Name
shugoshin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012409.3 Intron NP_001012409.1
NM_001012410.4 Intron NP_001012410.1
NM_001012411.3 Intron NP_001012411.1
NM_001012412.4 Intron NP_001012412.1
NM_001012413.3 Intron NP_001012413.1
NM_001199251.2 Intron NP_001186180.1
NM_001199252.2 Intron NP_001186181.1
NM_001199253.2 Intron NP_001186182.1
NM_001199254.2 Intron NP_001186183.1
NM_001199255.2 Intron NP_001186184.1
NM_001199256.2 Intron NP_001186185.1
NM_001199257.2 Intron NP_001186186.1
NM_138484.4 Intron NP_612493.1
XM_011533373.2 Intron XP_011531675.1
XM_011533375.2 Intron XP_011531677.1
XM_011533376.2 Intron XP_011531678.1
XM_011533377.2 Intron XP_011531679.1

View Full Product Details