Product Details

SNP ID
rs75272640
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:24117200 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTCATGTCCTGGCCAAATAAAAA[C/T]TTGGAGATGAGCTTCCTCTGTGTGA
Phenotype
MIM: 190160
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
THRB PubMed Links

Gene Details

Gene
THRB
Gene Name
thyroid hormone receptor beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000461.4 7492 UTR 3 NP_000452.2
NM_001128176.2 7492 UTR 3 NP_001121648.1
NM_001128177.1 7492 UTR 3 NP_001121649.1
NM_001252634.1 7492 UTR 3 NP_001239563.1
XM_005265421.4 7492 Intron XP_005265478.1
XM_005265423.4 7492 Intron XP_005265480.1
XM_005265424.3 7492 Intron XP_005265481.1
XM_006713317.3 7492 Intron XP_006713380.1
XM_006713318.3 7492 Intron XP_006713381.1
XM_011534046.2 7492 Intron XP_011532348.1
XM_011534047.2 7492 Intron XP_011532349.1
XM_011534048.2 7492 Intron XP_011532350.1
XM_011534049.2 7492 Intron XP_011532351.1
XM_011534050.2 7492 Intron XP_011532352.1
XM_011534051.2 7492 Intron XP_011532353.1
XM_011534052.2 7492 Intron XP_011532354.1
XM_011534053.2 7492 Intron XP_011532355.1
XM_017007108.1 7492 Intron XP_016862597.1
XM_017007109.1 7492 Intron XP_016862598.1
XM_017007110.1 7492 Intron XP_016862599.1
XM_017007111.1 7492 Intron XP_016862600.1
XM_017007112.1 7492 Intron XP_016862601.1
XM_017007113.1 7492 Intron XP_016862602.1
XM_017007114.1 7492 Intron XP_016862603.1
XM_017007115.1 7492 Intron XP_016862604.1

View Full Product Details