Product Details

SNP ID
rs78543378
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:24118544 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATGGGAAAAAAAAGGATTTTCTA[C/T]GAAGAAAGCATGGAGAACTAATTTG
Phenotype
MIM: 190160
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
THRB PubMed Links

Gene Details

Gene
THRB
Gene Name
thyroid hormone receptor beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000461.4 6148 UTR 3 NP_000452.2
NM_001128176.2 6148 UTR 3 NP_001121648.1
NM_001128177.1 6148 UTR 3 NP_001121649.1
NM_001252634.1 6148 UTR 3 NP_001239563.1
XM_005265421.4 6148 Intron XP_005265478.1
XM_005265423.4 6148 Intron XP_005265480.1
XM_005265424.3 6148 Intron XP_005265481.1
XM_006713317.3 6148 UTR 3 XP_006713380.1
XM_006713318.3 6148 Intron XP_006713381.1
XM_011534046.2 6148 Intron XP_011532348.1
XM_011534047.2 6148 Intron XP_011532349.1
XM_011534048.2 6148 Intron XP_011532350.1
XM_011534049.2 6148 Intron XP_011532351.1
XM_011534050.2 6148 Intron XP_011532352.1
XM_011534051.2 6148 Intron XP_011532353.1
XM_011534052.2 6148 Intron XP_011532354.1
XM_011534053.2 6148 Intron XP_011532355.1
XM_017007108.1 6148 Intron XP_016862597.1
XM_017007109.1 6148 Intron XP_016862598.1
XM_017007110.1 6148 Intron XP_016862599.1
XM_017007111.1 6148 Intron XP_016862600.1
XM_017007112.1 6148 Intron XP_016862601.1
XM_017007113.1 6148 Intron XP_016862602.1
XM_017007114.1 6148 Intron XP_016862603.1
XM_017007115.1 6148 Intron XP_016862604.1

View Full Product Details