Product Details

SNP ID
rs79270057
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:24120002 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCTCCCTGCCGAGGGGGTGGGAG[C/T]GCAGGTGTGAAGGGTATCTCCATCA
Phenotype
MIM: 190160
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
THRB PubMed Links

Gene Details

Gene
THRB
Gene Name
thyroid hormone receptor beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000461.4 4690 UTR 3 NP_000452.2
NM_001128176.2 4690 UTR 3 NP_001121648.1
NM_001128177.1 4690 UTR 3 NP_001121649.1
NM_001252634.1 4690 UTR 3 NP_001239563.1
XM_005265421.4 4690 UTR 3 XP_005265478.1
XM_005265423.4 4690 UTR 3 XP_005265480.1
XM_005265424.3 4690 UTR 3 XP_005265481.1
XM_006713317.3 4690 UTR 3 XP_006713380.1
XM_006713318.3 4690 UTR 3 XP_006713381.1
XM_011534046.2 4690 UTR 3 XP_011532348.1
XM_011534047.2 4690 UTR 3 XP_011532349.1
XM_011534048.2 4690 UTR 3 XP_011532350.1
XM_011534049.2 4690 UTR 3 XP_011532351.1
XM_011534050.2 4690 UTR 3 XP_011532352.1
XM_011534051.2 4690 UTR 3 XP_011532353.1
XM_011534052.2 4690 UTR 3 XP_011532354.1
XM_011534053.2 4690 UTR 3 XP_011532355.1
XM_017007108.1 4690 UTR 3 XP_016862597.1
XM_017007109.1 4690 UTR 3 XP_016862598.1
XM_017007110.1 4690 UTR 3 XP_016862599.1
XM_017007111.1 4690 UTR 3 XP_016862600.1
XM_017007112.1 4690 UTR 3 XP_016862601.1
XM_017007113.1 4690 UTR 3 XP_016862602.1
XM_017007114.1 4690 UTR 3 XP_016862603.1
XM_017007115.1 4690 UTR 3 XP_016862604.1

View Full Product Details