Product Details

SNP ID
rs74366044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:39055918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCAGCTAAATGCCAAGAGGACTT[C/T]TGTGCTCACTACCGTATCCTCAGCA
Phenotype
MIM: 604385 MIM: 612167
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SCN11A PubMed Links

Gene Details

Gene
SCN11A
Gene Name
sodium voltage-gated channel alpha subunit 11
There are no transcripts associated with this gene.

Gene
WDR48
Gene Name
WD repeat domain 48
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303402.1 Intron NP_001290331.1
NM_001303403.1 Intron NP_001290332.1
NM_020839.3 Intron NP_065890.1
XM_005265346.3 Intron XP_005265403.1
XM_011533971.1 Intron XP_011532273.1
XM_017006960.1 Intron XP_016862449.1

View Full Product Details