Product Details

SNP ID
rs77388437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:57274813 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCAATACAGAAACCATAAGCCAT[C/T]ATTCATTTGAAAACACTTGTAATTC
Phenotype
MIM: 604299
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APPL1 PubMed Links

Gene Details

Gene
APPL1
Gene Name
adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 1
There are no transcripts associated with this gene.

Gene
ASB14
Gene Name
ankyrin repeat and SOCS box containing 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142733.2 Intron NP_001136205.2
NM_130387.5 Intron NP_569058.1
XM_017005736.1 Intron XP_016861225.1
XM_017005737.1 Intron XP_016861226.1
Gene
LOC105377102
Gene Name
uncharacterized LOC105377102
There are no transcripts associated with this gene.

View Full Product Details