Product Details

SNP ID
rs75693443
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70154269 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTAGACAGACTAAAAAAGCCAT[G/T]TATTCTTTCGTTTCTCTCTAAAAGA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CSN1S2BP PubMed Links

Gene Details

Gene
CSN1S2BP
Gene Name
casein alpha s2-like B, pseudogene
There are no transcripts associated with this gene.

Gene
PRR27
Gene Name
proline rich 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_214711.3 83 UTR 5 NP_999876.2

View Full Product Details