Product Details

SNP ID
rs77399495
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:56435849 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAGTAGCGTAATGGGAGTAACGG[A/T]CTTAACCTCATTTCTCTTTCAGAGC
Phenotype
MIM: 172439 MIM: 172450
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC100506514 PubMed Links

Gene Details

Gene
LOC100506514
Gene Name
uncharacterized LOC100506514
There are no transcripts associated with this gene.

Gene
PAICS
Gene Name
phosphoribosylaminoimidazole carboxylase; phosphoribosylaminoimidazolesuccinocarboxamide synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079524.1 101 Intron NP_001072992.1
NM_001079525.1 101 Intron NP_001072993.1
NM_006452.3 101 UTR 5 NP_006443.1
XM_011534366.1 101 Intron XP_011532668.1
Gene
PPAT
Gene Name
phosphoribosyl pyrophosphate amidotransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002703.4 101 Intron NP_002694.3
XM_011534387.1 101 Intron XP_011532689.1

View Full Product Details