Product Details

SNP ID
rs79922429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:176392792 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCACCTAGCGGGACAGTGGCGT[C/T]TGCTTCAGGGACATGAGCACCGAGC
Phenotype
MIM: 118970 MIM: 612861
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARL10 PubMed Links

Gene Details

Gene
ARL10
Gene Name
ADP ribosylation factor like GTPase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317948.1 823 Intron NP_001304877.1
NM_173664.5 823 Intron NP_775935.1
XM_011534529.2 823 Intron XP_011532831.1
XM_011534530.2 823 Intron XP_011532832.1
XM_011534531.2 823 Intron XP_011532833.1
XM_017009372.1 823 Intron XP_016864861.1
Gene
CLTB
Gene Name
clathrin light chain B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001834.3 823 Silent Mutation CAA,CAG Q206Q NP_001825.1
NM_007097.3 823 Silent Mutation CAA,CAG Q224Q NP_009028.1
XM_017009026.1 823 Silent Mutation CAA,CAG Q139Q XP_016864515.1
XM_017009027.1 823 Intron XP_016864516.1
Gene
HIGD2A
Gene Name
HIG1 hypoxia inducible domain family member 2A
There are no transcripts associated with this gene.

Gene
NOP16
Gene Name
NOP16 nucleolar protein
There are no transcripts associated with this gene.

View Full Product Details