Product Details

SNP ID
rs77634146
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:176544221 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTTTAAAGCTGCCGTTTGTTGAC[A/G]GAACAGCCACCTAAAGGAGAGCTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDHR2 PubMed Links

Gene Details

Gene
CDHR2
Gene Name
cadherin related family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171976.1 Intron NP_001165447.1
NM_017675.4 Intron NP_060145.3
Gene
RNF44
Gene Name
ring finger protein 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014901.4 Intron NP_055716.1
XM_005265840.2 Intron XP_005265897.1
XM_005265841.4 Intron XP_005265898.1
XM_005265842.4 Intron XP_005265899.1
XM_005265843.2 Intron XP_005265900.1
XM_005265844.3 Intron XP_005265901.1
XM_005265845.4 Intron XP_005265902.1
XM_006714831.2 Intron XP_006714894.1
XM_006714832.2 Intron XP_006714895.1
XM_011534466.2 Intron XP_011532768.1
XM_011534467.2 Intron XP_011532769.1

View Full Product Details