Product Details

SNP ID
rs75029982
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:160423355 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTTAGGTGACAATTTGCTAGCA[C/T]GTTGATCTTCGTGTGAGTGTGAGTC
Phenotype
MIM: 604147 MIM: 605974
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTTG1 PubMed Links

Gene Details

Gene
PTTG1
Gene Name
pituitary tumor-transforming 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282382.1 Intron NP_001269311.1
NM_001282383.1 Intron NP_001269312.1
NM_004219.3 Intron NP_004210.1
Gene
SLU7
Gene Name
SLU7 homolog, splicing factor
There are no transcripts associated with this gene.

View Full Product Details