Product Details

SNP ID
rs80057678
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137176983 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTCTATCCAGGCAGCTCTGAGAC[A/G]TGGCCAAGGGTGTGGCTGGACCAGG
Phenotype
MIM: 606946 MIM: 138249 MIM: 613505
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANAPC2 PubMed Links

Gene Details

Gene
ANAPC2
Gene Name
anaphase promoting complex subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013366.3 Intron NP_037498.1
Gene
GRIN1
Gene Name
glutamate ionotropic receptor NMDA type subunit 1
There are no transcripts associated with this gene.

Gene
LRRC26
Gene Name
leucine rich repeat containing 26
There are no transcripts associated with this gene.

Gene
MIR3621
Gene Name
microRNA 3621
There are no transcripts associated with this gene.

Gene
TMEM210
Gene Name
transmembrane protein 210
There are no transcripts associated with this gene.

View Full Product Details