Product Details

SNP ID
rs76781099
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:36847728 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCAGGAGACTGGAACGTGGGAAA[C/T]GAGAAAGTGTAAGTCGACGTGTGAT
Phenotype
MIM: 603295
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SMAD9 PubMed Links

Gene Details

Gene
SMAD9
Gene Name
SMAD family member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127217.2 2584 UTR 3 NP_001120689.1
NM_005905.5 2584 UTR 3 NP_005896.1
XM_005266401.3 2584 Intron XP_005266458.1
XM_005266403.3 2584 Intron XP_005266460.1
XM_005266404.3 2584 Intron XP_005266461.1
XM_006719827.3 2584 Intron XP_006719890.1

View Full Product Details