Product Details

SNP ID
rs75126966
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:44385721 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTTTTGTCAGCTGAATGACCTTG[A/G]ACAAGGTATTTACTCTTGGTATTGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SERP2 PubMed Links

Gene Details

Gene
SERP2
Gene Name
stress-associated endoplasmic reticulum protein family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010897.1 Intron NP_001010897.1

View Full Product Details