Product Details

SNP ID
rs78856045
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:45395794 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATGTTAGTTTATGCCATTAAAC[A/G]TTTGATGAAGAGCATCCAATGCCAA
Phenotype
MIM: 610793
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC25A30 PubMed Links

Gene Details

Gene
SLC25A30
Gene Name
solute carrier family 25 member 30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010875.3 1415 UTR 3 NP_001010875.1
NM_001286806.1 1415 UTR 3 NP_001273735.1
NM_001286807.1 1415 UTR 3 NP_001273736.1
XM_005266321.2 1415 UTR 3 XP_005266378.1
XM_005266322.1 1415 Intron XP_005266379.1
XM_006719793.3 1415 Intron XP_006719856.1
XM_006719794.1 1415 Intron XP_006719857.1
XM_011535026.1 1415 Intron XP_011533328.1
XM_011535027.2 1415 UTR 3 XP_011533329.1
XM_017020523.1 1415 Intron XP_016876012.1
XM_017020524.1 1415 UTR 3 XP_016876013.1
Gene
TPT1-AS1
Gene Name
TPT1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details