Product Details

SNP ID
rs74434951
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:23471246 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGTTACACAAAGAACCTCTTAT[A/G]CCACACAAAGGAATTTGGATTTTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00327 PubMed Links

Gene Details

Gene
LINC00327
Gene Name
long intergenic non-protein coding RNA 327
There are no transcripts associated with this gene.

View Full Product Details