Product Details
- SNP ID
-
rs75863106
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:27945516 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGATCTCATTTCATGGGAAAAAAA[A/T]ATCCTGTCTTGTTCATAAATTGACA
- Phenotype
-
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ATP5EP2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs34588959] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATP5EP2
- Gene Name
- ATP synthase, H+ transporting, mitochondrial F1 complex, epsilon subunit pseudogene 2
There are no transcripts associated with this gene.
- Gene
- LINC00543
- Gene Name
- long intergenic non-protein coding RNA 543
There are no transcripts associated with this gene.
View Full Product Details