Product Details

SNP ID
rs79224538
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:109392967 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGGAGAAAAAAATGGAAAATTTA[A/G]TCAGTCATAAGTTTTCATATTTAGC
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PPIL6 PubMed Links

Gene Details

Gene
PPIL6
Gene Name
peptidylprolyl isomerase like 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111298.2 Intron NP_001104768.2
NM_001286360.1 Intron NP_001273289.1
NM_001286361.1 Intron NP_001273290.1
NM_173672.4 Intron NP_775943.1
XM_011535765.2 Intron XP_011534067.1
XM_011535766.2 Intron XP_011534068.1
XM_011535767.2 Intron XP_011534069.1
XM_011535769.2 Intron XP_011534071.1
XM_017010774.1 Intron XP_016866263.1

View Full Product Details