Product Details

SNP ID
rs80057960
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:110614286 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGAAGTGCTGGGATTAGATCAGA[C/T]GCTTTATCAGAGAAGTCACAATGGA
Phenotype
MIM: 614720
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDK19 PubMed Links

Gene Details

Gene
CDK19
Gene Name
cyclin dependent kinase 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300960.1 1521 UTR 3 NP_001287889.1
NM_001300963.1 1521 UTR 3 NP_001287892.1
NM_001300964.1 1521 UTR 3 NP_001287893.1
NM_015076.4 1521 UTR 3 NP_055891.1
XM_005266871.3 1521 UTR 3 XP_005266928.1
XM_011535630.2 1521 UTR 3 XP_011533932.1
XM_011535631.2 1521 UTR 3 XP_011533933.1
XM_011535632.1 1521 Intron XP_011533934.1
XM_017010587.1 1521 UTR 3 XP_016866076.1
XM_017010588.1 1521 UTR 3 XP_016866077.1

View Full Product Details