Product Details
- SNP ID
-
rs77580112
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:111499944 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACAACACAAAACAAAACAAAAACT[A/G]CCCTAATTGACATTGTATGTGATAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TRAF3IP2-AS1
PubMed Links
Gene Details
- Gene
- TRAF3IP2-AS1
- Gene Name
- TRAF3IP2 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details