Product Details
- SNP ID
-
rs76039645
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:99609272 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACTCCTTGGCTCAGTGGTTCGACA[C/T]CCCCACCACAGCGACTCCGACTCAC
- Phenotype
-
MIM: 616741
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PRDM13
PubMed Links
Gene Details
- Gene
- PRDM13
- Gene Name
- PR domain 13
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021620.3 |
623 |
Missense Mutation |
ACC,ATC |
T121I |
NP_067633.2 |
View Full Product Details