Product Details

SNP ID
rs74728933
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:100389229 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAAACAATAAGGAAAATAAACAT[G/T]TTGCCTATTCCTTCAGAGTACTGCT
Phenotype
MIM: 603128
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SIM1 PubMed Links

Gene Details

Gene
SIM1
Gene Name
single-minded family bHLH transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005068.2 4216 UTR 3 NP_005059.2
XM_005267100.2 4216 UTR 3 XP_005267157.1
XM_011536072.2 4216 Intron XP_011534374.1
XM_017011197.1 4216 UTR 3 XP_016866686.1

View Full Product Details