Product Details

SNP ID
rs75724670
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:149920934 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGTCCTTCAAGAACAAGGCTGC[C/T]TTGCCTAATTTGCAGAATAAAGCCA
Phenotype
MIM: 609244
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAET1E-AS1 PubMed Links

Gene Details

Gene
RAET1E-AS1
Gene Name
RAET1E antisense RNA 1
There are no transcripts associated with this gene.

Gene
RAET1G
Gene Name
retinoic acid early transcript 1G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001788.3 Intron NP_001001788.2
XM_011535800.2 Intron XP_011534102.1
XM_017010831.1 Intron XP_016866320.1

View Full Product Details