Product Details

SNP ID
rs78244842
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:154410346 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTTGCTTGGATGAAACGTACCTT[C/T]CCTGCCATGACCTCGAGGTCTGGCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNKSR3 PubMed Links

Gene Details

Gene
CNKSR3
Gene Name
CNKSR family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173515.2 4354 Missense Mutation AAA,GAA K456E NP_775786.2
XM_011535485.2 4354 Missense Mutation AAA,GAA K446E XP_011533787.1

View Full Product Details