Product Details

SNP ID
rs77488359
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:166931833 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGCCCCTTGCCTCAGACACTGTC[C/T]ACTCCTTCCACGAAGCTCTCCTGTG
Phenotype
MIM: 612944
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNASET2 PubMed Links

Gene Details

Gene
RNASET2
Gene Name
ribonuclease T2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003730.4 Intron NP_003721.2
XM_017011397.1 Intron XP_016866886.1
XM_017011398.1 Intron XP_016866887.1
XM_017011399.1 Intron XP_016866888.1

View Full Product Details