Product Details

SNP ID
rs74583382
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:168293355 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATATTAAATTTTCACATCATTTT[G/T]CAATAATATATATGCACGTAAACTG
Phenotype
MIM: 608966
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DACT2 PubMed Links

Gene Details

Gene
DACT2
Gene Name
dishevelled binding antagonist of beta catenin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286350.1 1503 Intron NP_001273279.1
NM_001286351.1 1503 UTR 3 NP_001273280.1
NM_214462.4 1503 Intron NP_999627.2
XM_011535507.2 1503 Intron XP_011533809.1

View Full Product Details