Product Details

SNP ID
rs77739229
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:39149365 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCAAAAGGGTAATAAATAAATGG[A/G]AAACAAAGTATAGTTTTAGTAATCA
Phenotype
MIM: 610397
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRAPPC6B PubMed Links

Gene Details

Gene
TRAPPC6B
Gene Name
trafficking protein particle complex 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079537.1 1797 UTR 3 NP_001073005.1
NM_177452.3 1797 UTR 3 NP_803235.1
XM_011536409.2 1797 UTR 3 XP_011534711.1
XM_017020964.1 1797 Intron XP_016876453.1

View Full Product Details