Product Details

SNP ID
rs77465149
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:39149642 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTCCATGCACGGGGAGAGAAGTT[A/G]AAAGTCAAAACTGTAACAGGAATAA
Phenotype
MIM: 610397
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRAPPC6B PubMed Links

Gene Details

Gene
TRAPPC6B
Gene Name
trafficking protein particle complex 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079537.1 1520 UTR 3 NP_001073005.1
NM_177452.3 1520 UTR 3 NP_803235.1
XM_011536409.2 1520 UTR 3 XP_011534711.1
XM_017020964.1 1520 Intron XP_016876453.1

View Full Product Details