Product Details

SNP ID
rs76134963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:45095209 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGAAGATATTTCTCTTTTTTTC[G/T]TGTTTCCACAGATCGTTAACTGAAG
Phenotype
MIM: 614907
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PRPF39 PubMed Links

Gene Details

Gene
PRPF39
Gene Name
pre-mRNA processing factor 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017922.3 Intron NP_060392.3

View Full Product Details