Product Details

SNP ID
rs75728940
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:45128926 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACCTTAGTAGCTACTCAATAGAT[A/G]TAACATTTATTAAATATTAATTGTG
Phenotype
MIM: 609644 MIM: 186947
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FANCM PubMed Links

Gene Details

Gene
FANCM
Gene Name
Fanconi anemia complementation group M
There are no transcripts associated with this gene.

Gene
FKBP3
Gene Name
FK506 binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002013.3 Intron NP_002004.1
XM_011536565.2 Intron XP_011534867.1
XM_017021092.1 Intron XP_016876581.1
XM_017021093.1 Intron XP_016876582.1

View Full Product Details