Product Details

SNP ID
rs74955576
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:60245953 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGAGAAGGGCAAAGAAGGCAAAA[A/C]GAGAAGAGAAGAAAAAGGGAGGAGA
Phenotype
MIM: 606108
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC101927702 PubMed Links

Gene Details

Gene
LOC101927702
Gene Name
uncharacterized LOC101927702
There are no transcripts associated with this gene.

Gene
PPM1A
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021003.4 197 Intron NP_066283.1
NM_177951.2 197 Intron NP_808820.1
NM_177952.2 197 Silent Mutation AGA,CGA R36R NP_808821.2
XM_005267779.1 197 UTR 5 XP_005267836.1
XM_005267781.1 197 Intron XP_005267838.1
XM_011536872.1 197 Intron XP_011535174.1
XM_011536876.1 197 Intron XP_011535178.1
XM_011536877.1 197 Intron XP_011535179.1
XM_011536878.1 197 Intron XP_011535180.1
XM_011536879.2 197 Intron XP_011535181.1
XM_011536880.2 197 Intron XP_011535182.1
XM_011536881.2 197 Intron XP_011535183.1
XM_011536882.2 197 Intron XP_011535184.1
XM_011536883.1 197 Intron XP_011535185.1
XM_011536884.1 197 Intron XP_011535186.1
XM_017021381.1 197 Intron XP_016876870.1
XM_017021382.1 197 Intron XP_016876871.1
XM_017021383.1 197 Intron XP_016876872.1
XM_017021384.1 197 Intron XP_016876873.1
XM_017021385.1 197 Intron XP_016876874.1
XM_017021386.1 197 Intron XP_016876875.1
XM_017021387.1 197 Intron XP_016876876.1

View Full Product Details