Product Details
- SNP ID
-
rs77147457
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:23939470 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAATCATGGTGCATAAGTATTTAT[A/G]GTTTCTTACAAAGTATGAAAAGGTA
- Phenotype
-
MIM: 616925
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DHRS4-AS1
PubMed Links
Gene Details
- Gene
- DHRS4-AS1
- Gene Name
- DHRS4 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details