Product Details

SNP ID
rs77528519
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24096678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTCATTTGGCCATATCTGTCCCT[C/T]GCTGAGCAGGAGACACCCTCCTCAA
Phenotype
MIM: 162080 MIM: 614095
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NRL PubMed Links

Gene Details

Gene
NRL
Gene Name
neural retina leucine zipper
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006177.3 Intron NP_006168.1
XM_005267708.4 Intron XP_005267765.1
XM_005267709.3 Intron XP_005267766.1
XM_005267710.3 Intron XP_005267767.1
XM_011536801.2 Intron XP_011535103.2
XM_011536802.1 Intron XP_011535104.1
XM_011536804.2 Intron XP_011535106.1
XM_011536805.2 Intron XP_011535107.1
XM_011536806.2 Intron XP_011535108.2
XM_017021350.1 Intron XP_016876839.1
Gene
PCK2
Gene Name
phosphoenolpyruvate carboxykinase 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018073.2 Intron NP_001018083.2
NM_001291556.1 Intron NP_001278485.1
NM_001308054.1 Intron NP_001294983.1
NM_004563.3 Intron NP_004554.3
XM_006720158.2 Intron XP_006720221.1

View Full Product Details