Product Details

SNP ID
rs78265092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:75083517 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGGAAAAATGATACAGATTCATT[C/T]GTTACCACATGGCTTCATAAAACAC
Phenotype
MIM: 609798
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NEK9 PubMed Links

Gene Details

Gene
NEK9
Gene Name
NIMA related kinase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005268208.1 3961 UTR 3 XP_005268265.1
XM_005268209.3 3961 UTR 3 XP_005268266.1
XM_017021765.1 3961 UTR 3 XP_016877254.1
Gene
ZC2HC1C
Gene Name
zinc finger C2HC-type containing 1C
There are no transcripts associated with this gene.

View Full Product Details