Product Details

SNP ID
rs78655524
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:96363990 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGTTTGCAGAGCGCTTCTCGCA[G/T]ACGCTCGCTTAATCCTGGGTGTGTC
Phenotype
MIM: 616226 MIM: 616605
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ATG2B PubMed Links

Gene Details

Gene
ATG2B
Gene Name
autophagy related 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006720187.2 Intron XP_006720250.1
XM_017021397.1 Intron XP_016876886.1
Gene
GSKIP
Gene Name
GSK3B interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271904.1 Intron NP_001258833.1
NM_001271905.1 Intron NP_001258834.1
NM_001271906.1 Intron NP_001258835.1
NM_016472.4 Intron NP_057556.2
XM_017021365.1 Intron XP_016876854.1

View Full Product Details