Product Details
- SNP ID
-
rs76690622
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:103916164 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCACTTCATAATAACTGCCAATTA[A/T]GATAGAACTTATTTAATCAATAAAG
- Phenotype
-
MIM: 604573
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C14orf2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs199787171] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C14orf2
- Gene Name
- chromosome 14 open reading frame 2
View Full Product Details