Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000462.3 | 3040 | Silent Mutation | TGA,TTA | *876L | NP_000453.2 |
NM_130838.1 | 3040 | Silent Mutation | TGA,TTA | *853L | NP_570853.1 |
NM_130839.2 | 3040 | Silent Mutation | TGA,TTA | *873L | NP_570854.1 |
XM_005268267.4 | 3040 | Silent Mutation | XP_005268324.1 | ||
XM_005268268.4 | 3040 | Silent Mutation | XP_005268325.1 | ||
XM_005268269.4 | 3040 | Silent Mutation | XP_005268326.1 | ||
XM_005268270.4 | 3040 | Silent Mutation | XP_005268327.1 | ||
XM_005268271.4 | 3040 | Silent Mutation | XP_005268328.1 | ||
XM_006720675.3 | 3040 | Silent Mutation | XP_006720738.1 | ||
XM_006720676.3 | 3040 | Silent Mutation | XP_006720739.1 | ||
XM_011521994.2 | 3040 | Silent Mutation | XP_011520296.1 | ||
XM_011521995.2 | 3040 | Silent Mutation | XP_011520297.1 | ||
XM_017022544.1 | 3040 | Silent Mutation | XP_016878033.1 | ||
XM_017022545.1 | 3040 | Silent Mutation | XP_016878034.1 | ||
XM_017022546.1 | 3040 | Silent Mutation | XP_016878035.1 | ||
XM_017022547.1 | 3040 | Silent Mutation | XP_016878036.1 | ||
XM_017022548.1 | 3040 | Silent Mutation | XP_016878037.1 | ||
XM_017022549.1 | 3040 | Silent Mutation | XP_016878038.1 | ||
XM_017022550.1 | 3040 | Silent Mutation | XP_016878039.1 | ||
XM_017022551.1 | 3040 | Silent Mutation | XP_016878040.1 | ||
XM_017022552.1 | 3040 | Silent Mutation | XP_016878041.1 | ||
XM_017022553.1 | 3040 | Silent Mutation | XP_016878042.1 | ||
XM_017022554.1 | 3040 | Silent Mutation | XP_016878043.1 | ||
XM_017022555.1 | 3040 | Silent Mutation | XP_016878044.1 | ||
XM_017022556.1 | 3040 | Silent Mutation | XP_016878045.1 | ||
XM_017022557.1 | 3040 | Silent Mutation | XP_016878046.1 | ||
XM_017022558.1 | 3040 | Silent Mutation | XP_016878047.1 | ||
XM_017022559.1 | 3040 | Silent Mutation | XP_016878048.1 |